Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-18-3 | C2H6S | 62.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA127760010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Acidity=<0.0002 meq/g
UVat 210 nm A: =<0.699
UVat 245 nm A: =<0.046
Residue after evaporation=<0.0003 %