alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
CAS: 98-83-9 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA127711000
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2206-27-1
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.02 % (Coulometric)
D-Enrichment>=99.95 %
Appearance (Color)Clear colorless