2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals
2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals
2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals
2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals
CAS: 107-83-5 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA127341001
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1712-64-7
IUPAC Namepropan-2-yl nitrate
Molecular FormulaC3H7NO3
InChI KeyGAPFWGOSHOCNBM-UHFFFAOYSA-N
SMILESCC(C)O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%