1-Methylimidazole, 99%, Thermo Scientific Chemicals
1-Methylimidazole, 99%, Thermo Scientific Chemicals
1-Methylimidazole, 99%, Thermo Scientific Chemicals
1-Methylimidazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methylimidazole, 99%, Thermo Scientific Chemicals
CAS: 616-47-7 | C4H6N2 | 82.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA126995000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS126-33-0
IUPAC Name1λ⁶-thiolane-1,1-dione
Molecular FormulaC4H8O2S
InChI KeyHXJUTPCZVOIRIF-UHFFFAOYSA-N
SMILESO=S1(=O)CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.3 %
Appearance (Form)Liquid after melting
Refractive index1.4820 to 1.4850 (20°C, 589 nm)
Appearance (Color)Clear yellow