2-Methylheptane, 99%, Thermo Scientific Chemicals
2-Methylheptane, 99%, Thermo Scientific Chemicals
2-Methylheptane, 99%, Thermo Scientific Chemicals
2-Methylheptane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methylheptane, 99%, Thermo Scientific Chemicals
CAS: 592-27-8 | C8H18 | 114.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA126880250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-99-3
IUPAC Namebenzene-1,2-dicarboxylic acid
Molecular FormulaC8H6O4
InChI KeyXNGIFLGASWRNHJ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point200°C to 205°C (Dec)
Titration with NaOH>=98.5 %
Sulfate (SO4)=<50 ppm
Iron (Fe)=<50 ppm