p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
CAS: 830-09-1 | C10H10O3 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA125841000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-07-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=97.5 %
Water=<0.2 %
Melting point116°C to 120°C
Ammonium chloride (NH4Cl)=<0.5 %