3-Mercaptopropionic acid, 99%, Thermo Scientific Chemicals
3-Mercaptopropionic acid, 99%, Thermo Scientific Chemicals
3-Mercaptopropionic acid, 99%, Thermo Scientific Chemicals
3-Mercaptopropionic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Mercaptopropionic acid, 99%, Thermo Scientific Chemicals
CAS: 107-96-0 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA125535000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material3-Mercaptopropionic acid
CAS107-96-0
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Reduction reagent
RUO – Research Use Only