Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
CAS: 68-11-1 | C2H4O2S | 92.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA125430100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-67-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.3704 to 1.3724 (20°C, 589 nm)