Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
CAS: 68-11-1 | C2H4O2S | 92.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
250 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA125430010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMercaptoacetic acid
CAS68-11-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre