DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
CAS: 90-64-2 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Plastic bottle
Catalog number FSA125312500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS8013-01-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown
Assay (special)Total aerobic microbial count: ≤ 5000 cfu/g
pH6.5-7.5
FormPowder
Assay (special)Sulfuric ashes: ≤ 17%