Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number FSA125240250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS122-04-3
IUPAC Name4-nitrobenzoyl chloride
Molecular FormulaC7H4ClNO3
InChI KeySKDHHIUENRGTHK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystalline powder or crystals or flakes
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)70-75?C