Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
CAS: 110-16-7 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
View all
Catalog number FSA125231000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS164149-28-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)214.0-220.0?C