Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number FSA124210025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS373-57-9
IUPAC Nametrifluoroborane
Molecular FormulaBF3
InChI KeyWTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILESFB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to amber
Appearance (Form)Solution
Assay10 to 15 % w/v (BF3)
Volumetric standard solution
RUO – Research Use Only