Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA123990010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS3811-04-9
IUPAC Namepotassium chlorate
Molecular FormulaClKO3
InChI KeyVKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration>=99.0 %
Insoluble matter=<0.005 %
Bromate (BrO3)=<150 ppm
Sodium (Na)=<100 ppm
Heavy metals (as Pb)=<5 ppm