Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA123170010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS65-82-7
IUPAC Name(2S)-2-acetamido-4-(methylsulfanyl)butanoic acid
Molecular FormulaC7H13NO3S
InChI KeyXUYPXLNMDZIRQH-LURJTMIESA-N
SMILESCSCC[C@H](NC(C)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)101.0-107.0?C
Optical Rotation-21.9 ± 0.1? (c=1.3 in water)
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
C-X bond formation reagent
RUO – Research Use Only