Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
CAS: 79-31-2 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
500 g
Packaging:
Glass bottle
Catalog number FSA122520010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer70 to 120 ppm Amylene
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.0 %
Water=<0.05 %
View more
Acylation reagent
RUO – Research Use Only