2-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
CAS: 614-75-5 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA121690100
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS65-71-4
IUPAC Name5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC5H6N2O2
InChI KeyRWQNBRDOKXIBIV-UHFFFAOYSA-N
SMILESCC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Powder
HPLC>=98.5 %