5-Hydroxy-2-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
5-Hydroxy-2-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
5-Hydroxy-2-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
5-Hydroxy-2-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
5-Hydroxy-2-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA121650050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS546-93-0
IUPAC Namemagnesium(2+) carbonate
Molecular FormulaCMgO3
InChI KeyZLNQQNXFFQJAID-UHFFFAOYSA-L
SMILES[Mg++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration excess HCl40.0 to 43.5 % (MgO)
Additional infoAbsence-specified microorganisms: Passes
IdentificationConforms