4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
CAS: 121-34-6 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA121351000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1732-09-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4325-1.4355 @ 20?C