4-Hydroxycoumarin, 98%, Thermo Scientific Chemicals
4-Hydroxycoumarin, 98%, Thermo Scientific Chemicals
4-Hydroxycoumarin, 98%, Thermo Scientific Chemicals
4-Hydroxycoumarin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxycoumarin, 98%, Thermo Scientific Chemicals
CAS: 1076-38-6 | C9H6O3 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA121105000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS14593-46-5
IUPAC Namesodium 2,2-dimethylpropan-1-olate
Molecular FormulaC5H11NaO
InChI KeyYQMLGVNRTAQUFQ-UHFFFAOYSA-N
SMILES[Na+].CC(C)(C)C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Solution
Titration with HClO4>=30 %
Appearance (Color)Yellow to orange to brown