p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
CAS: 501-98-4 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA121091000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7681-52-9
IUPAC Namesodium hypochlorite
Molecular FormulaClNaO
InChI KeySUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES[Na+].[O-]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Solution
Titration Iodometric4.0 to 6.0 %