Hexamethylbenzene, 98+%, Thermo Scientific Chemicals
Hexamethylbenzene, 98+%, Thermo Scientific Chemicals
Hexamethylbenzene, 98+%, Thermo Scientific Chemicals
Hexamethylbenzene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexamethylbenzene, 98+%, Thermo Scientific Chemicals
CAS: 87-85-4 | C12H18 | 162.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA120570050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.127 mm (0.005 in.) thick
Impurity contentTantalum: ≤0.06%
Total Metal Impurities≤0.03% (excluding Ta)