Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
CAS: 687-47-8 | C5H10O3 | 118.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA118662500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl L(-)-lactate
Acidity0.1% max. (as lactic acid)
CAS687-47-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.