Ethylenediaminetetraacetic acid, 99%, pure
Ethylenediaminetetraacetic acid, 99%, pure
Ethylenediaminetetraacetic acid, 99%, pure
Ethylenediaminetetraacetic acid, 99%, pure
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, 99%, pure
CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number FSA118430010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid
CAS60-00-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cau