Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals
Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals
Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals
Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number FSA118085000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS768-33-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.5065-1.5105 @ 20?C