Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
CAS: 140-88-5 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA117980010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2807-30-9
IUPAC Name2-propoxyethan-1-ol
Molecular FormulaC5H12O2
InChI KeyYEYKMVJDLWJFOA-UHFFFAOYSA-N
SMILESCCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.1 % (K.F.)