1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
Packaging:
View all
Metal Drum
Catalog number FSA117110250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.97% (metals basis)
Appearance (Color)Gray
FormRod, 5mm (0.2in) diameter.