5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
CAS: 77-71-4 | C5H8N2O2 | 128.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA116312500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2131-61-5
IUPAC Name1-isothiocyanato-4-nitrobenzene
Molecular FormulaC7H4N2O2S
InChI KeyNXHSSIGRWJENBH-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Melting Point (clear melt)106-112?C
FormPowder or lumpy solid
Appearance (Color)Cream to yellow
Assay (GC)≥96.0%