2,3-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
2,3-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
2,3-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
2,3-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
CAS: 86-51-1 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA115370100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 3-Dimethoxybenzaldehyde
Name Note97%
CAS86-51-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning