N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA114421000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS141-05-9
IUPAC Name1,4-diethyl (2Z)-but-2-enedioate
Molecular FormulaC8H12O4
InChI KeyIEPRKVQEAMIZSS-WAYWQWQTSA-N
SMILESCCOC(=O)C=C/C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Identification (FTIR)Conforms
Refractive Index1.4380-1.4430 @ 20?C
Assay (GC)≥96.0%