1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
CAS: 105-05-5 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA114150250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12125-01-8
IUPAC Nameamine dihydrofluoride
Molecular FormulaF2H5N
InChI KeyKVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILESN.F.F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0%