N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals
N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals
N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals
N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA113911000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS573-97-7
IUPAC Name1-bromonaphthalen-2-ol
Molecular FormulaC10H7BrO
InChI KeyFQJZPYXGPYJJIH-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Cream to pale brown
FormPowder
Melting Point (clear melt)73.0-83.0?C