2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
CAS: 94-75-7 | C8H6Cl2O3 | 221.033 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Catalog number FSA113631000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)61 to 71 %Ni
Appearance (Color)Black