3,5-Dichlorophenol, 99%, Thermo Scientific Chemicals
3,5-Dichlorophenol, 99%, Thermo Scientific Chemicals
3,5-Dichlorophenol, 99%, Thermo Scientific Chemicals
3,5-Dichlorophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dichlorophenol, 99%, Thermo Scientific Chemicals
CAS: 591-35-5 | C6H4Cl2O | 163 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA113610100
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 5-Dichlorophenol
CAS591-35-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0