1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
CAS: 4101-68-2 | C10H20Br2 | 300.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA112785000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26348-70-9
IUPAC Namedihydrogen methyl (2S)-2,6-diaminohexanoate dichloride
Molecular FormulaC7H18Cl2N2O2
InChI KeySXZCBVCQHOJXDR-ILKKLZGPSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Optical Rotation20.0 ± 1.5? (C=1 in methanol)
Assay (Titration ex Chloride)≥98.5 to ≤101.5%