1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
CAS: 280-57-9 | C6H12N2 | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass Bottle
Catalog number FSA112471000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS58966-93-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Appearance (Form)Crystalline powder
Titration Argentometric>=96.0 %