1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA111171000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.5380 to 1.5410 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %