4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
CAS: 1073-67-2 | C8H7Cl | 138.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA110090100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS64704-23-0
SpecificationsSpecification SheetSpecification Sheet
pH10.0-11.0 (1% in water)
Appearance (Color)White to to pale yellow-brown to pale brown
FormPowder or crystalline powder
Assay from Supplier's CofA≥98.0%
Loss on Drying≤2.0%