2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
CAS: 156-41-2 | C8H11ClN | 156.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA109880250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
CommentMay contain up to 10% methanol
Appearance (Color)Clear colorless to pale yellow
Assay (Titration)≥48.0 to ≤52.0%