4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
CAS: 604-44-4 | C10H7ClO | 178.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Catalog number FSA109551000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS696-42-4
IUPAC Name5-fluoropyridine-3-carbonitrile
Molecular FormulaC6H3FN2
InChI KeyLQJWCPZRNHAVJD-UHFFFAOYSA-N
SMILESFC1=CN=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps or fused solid
Melting Point (clear melt)47.0-57.0?C
Assay (GC)≥96.0%
Appearance (Color)Dark brown