2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
Packaging:
Catalog number FSA109500250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS14901-07-6
IUPAC Name(3E)-4-(2,6,6-trimethylcyclohex-1-en-1-yl)but-3-en-2-one
Molecular FormulaC13H20O
InChI KeyPSQYTAPXSHCGMF-BQYQJAHWSA-N
SMILESCC(=O)\C=C\C1=C(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95 %
Specific gravity(25°C) 0.941 to 0.947