4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA109371000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10025-91-9
IUPAC Nameantimony(3+) trichloride
Molecular FormulaCl3Sb
InChI KeyFAPDDOBMIUGHIN-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% SbCl₃ min
Impurity contentCa 0.005%
Impurity contentFe 0.002%
Impurity contentK 0.01%
Impurity contentPb 0.005%