4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 88-04-0 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA109065000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS536-60-7
IUPAC Name[4-(propan-2-yl)phenyl]methanol
Molecular FormulaC10H14O
InChI KeyOIGWAXDAPKFNCQ-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5180 to 1.5210 (20°C, 589 nm)