3-Chlorobenzoic acid, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoic acid, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoic acid, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chlorobenzoic acid, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA108665000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1641-09-4
IUPAC Namethiophene-3-carbonitrile
Molecular FormulaC5H3NS
InChI KeyGSXCEVHRIVLFJV-UHFFFAOYSA-N
SMILESN#CC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥97.5% (US-sourced material)
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5% (UK-sourced material)
CommentMaterial sourced in UK and US