tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
CAS: 870-46-2 | C5H12N2O2 | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA107880250
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS645-13-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5205-1.5255 @ 20?C