4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA107420050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3094-87-9
IUPAC Nameλ²-iron(2+) diacetate
Molecular FormulaC4H6FeO4
InChI KeyLNOZJRCUHSPCDZ-UHFFFAOYSA-L
SMILES[Fe++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)91% min