1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
CAS: 637-59-2 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Catalog number FSA107291000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS583-06-2
IUPAC Name(2E)-4-oxo-4-phenylbut-2-enoic acid
Molecular FormulaC10H8O3
InChI KeyPLPDHGOODMBBGN-VOTSOKGWSA-N
SMILESOC(=O)C=CC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)89.0-99.0?C
Appearance (Color)Yellow
Assay (GC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals long description.
RUO – Research Use Only