1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
CAS: 637-59-2 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA107290050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18063-02-0
IUPAC Name2,6-difluorobenzoyl chloride
Molecular FormulaC7H3ClF2O
InChI KeyQRHUZEVERIHEPT-UHFFFAOYSA-N
SMILESFC1=CC=CC(F)=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4970-1.5015 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow