3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Catalog number FSA107000250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS9002-93-1
IUPAC Name2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
InChI KeyJYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILESCC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Synonymtriton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy
Cloud point53.0 to 67.0? (1% inwater)
Water Content (Karl Fischer Titration)≤ 0.50%
FormViscous liquid
Chromatographic Purity53.0 to 67.0? (1% inwater)