2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 1072-85-1 | C6H4BrF | 175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number FSA106990100
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12150-46-8
IUPAC Nameλ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide)
Molecular FormulaC34H28FeP2
InChI KeyKZPYGQFFRCFCPP-UHFFFAOYSA-N
SMILES[Fe++].C1=C[C-](C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=C[C-](C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥96.0% (Phosphorus NMR)
Appearance (Color)Yellow to orange
FormCrystalline powder or powder
Proton NMRConforms to structure